Diphenylacetonitrile 99+%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2,2-Diphenylacetonitrile
150361000
150365000
150360025
ACRO150361000EA
43.4
EUR
ACRO150361000
ACRO150365000
ACRO150360025
Diphenylacetonitrile 99+%
Diphenylacetonitrile
Formula:
(C₆H₅)₂CHCN MW: 193.25 g/mol Boiling Pt: 181 °C (12 mmHg) Melting Pt: 73…76 °C Flash Pt: 120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001862 CAS Number: 86-29-3 EINECS: 201-662-5 Merck Index: 13,03348 |
Specification Test Results
Appearance | White to cream to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 73 to 76 °C |
Water | ≤0.2 % (K.F.) |
Sulfated ash | ≤0.1 % |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...