Diphenylacetonitrile ≥99%

Supplier: Thermo Fisher Scientific
Warning

L02404.22 L02404.36
L02404.22EA 36.3 EUR
L02404.22 L02404.36
Diphenylacetonitrile ≥99%
Diphenylacetonitrile

Purity: 99% CAS No.: 86-29-3 Molecular Formula: C14H11N Formula Weight: 193.25 Flash Point: 120°(248°F) Melting Point: 72-76° Boiling Point: 181°/12mm Beilstein Reference: 1911160 Merck Reference: 14,3316 MDL No.: MFCD00001862

Formula: (C₆H₅)₂CHCN
Boiling Pt: 181 °C (12 mmHg)
Melting Pt: 72…76 °C
Density: 1,182 g/cm³ (20 °C)
Flash Pt: 120 °C (248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001862
CAS Number: 86-29-3
EINECS: 201-662-5
Merck Index: 14,3316

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR