m-Xylene
Supplier: Roth Carl
|
Danger
|
3791.1
3791.2
ROTH3791.1EA
0
EUR
ROTH3791.1
ROTH3791.2
m-Xylene
m-Xylene
Formula:
C₈H₁₀ Boiling Pt: 138 °C (1013 hPa) Melting Pt: –48 °C Density: 0,864 g/cm³ (20 °C) Flash Pt: 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008536 CAS Number: 108-38-3 EINECS: 203-576-3 UN: 1307 ADR: 3,III Merck Index: 12,10214 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...