Cyclopropanecarbonitrile ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Cyclopropyl cyanide
A11125.09
A11125.18
A11125.30
A11125.09EA
42.8
EUR
A11125.09
A11125.18
A11125.30
Cyclopropanecarbonitrile ≥98%
Cyclopropanecarbonitrile
Purity: 98%
CAS No.: 5500-21-0
Molecular Formula: C4H5N
Formula Weight: 67.09
Flash Point: 32°(89°F)
Refractive Index: 1.4210
Odour: Recognizable
Boiling Point: 134-135°
Beilstein Reference: 635790
UN No.: 3275
MDL No.: MFCD00001269
Formula:
C₄H₅N MW: 67,09 g/mol Boiling Pt: 134…135 °C Melting Pt: –25 °C Density: 0,905 Flash Pt: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001269 CAS Number: 5500-21-0 EINECS: 226-836-8 UN: 3275 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...