Isophorone ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 3,5,5-Trimethyl-2-cyclohexen-1-one, 3,5,5-Trimethylcyclohex-2-enone

A14844.0F A14844.AK A14844.AP
A14844.0FEA 101 EUR
A14844.0F A14844.AK A14844.AP
Isophorone ≥97%
Isophorone

Purity: 97% CAS No.: 78-59-1 Molecular Formula: C9H14O Formula Weight: 138.21 Flash Point: 84°(183°F) Refractive Index: 1.4780 Melting Point: -8° Boiling Point: 99°/18mm Beilstein Reference: 1280721 Merck Reference: 14,5196 MDL No.: MFCD00001584

Formula: C₉H₁₄O
MW: 138,21 g/mol
Boiling Pt: 99 °C (18 mmHg)
Melting Pt: –8 °C
Density: 0,922
Flash Pt: 84 °C (183 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001584
CAS Number: 78-59-1
EINECS: 201-126-0
Merck Index: 14,5196

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR