Xanthone ≥99%
Supplier: Thermo Scientific
Synonyms:
Xanthen-9-one, 9H-Xanthen-9-one
A14812.14
A14812.22
A14812.14EA
49.7
EUR
A14812.14
A14812.22
Xanthone ≥99%
Xanthone
Purity: 99%
CAS No.: 90-47-1
Molecular Formula: C13H8O2
Formula Weight: 196.21
Melting Point: 174-178°
Boiling Point: 350-351°
Merck Reference: 14,10062
MDL No.: MFCD00005060
Formula:
C₁₃H₈O₂ MW: 196,21 g/mol Boiling Pt: 350…351 °C Melting Pt: 174…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005060 CAS Number: 90-47-1 EINECS: 201-997-7 Merck Index: 14,10062 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...