Xanthydrol, Supelco®

Supplier: Merck

Synonyms: 9-Hydroxyxanthene, 9-Xanthenol, 9H-Xanthen-9-ol

M-Clarity™ quality level = MQ100

95580-100G 95580-25G
95580-100GEA 704 EUR
95580-100G 95580-25G
Xanthydrol, Supelco®
Xanthydrol
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 338
Melting Pt: 123
MDL Number: MFCD00005057
CAS Number: 90-46-0
EINECS: 201-996-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6e2cc14de91278ac05209754aed751428631d84&vid=a512884c8f1880b8fd0940c2582a19dfb9d9fd63&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b4786c6b5ca5b97d6295c8fa78dcdcd621ae7b4&vid=bf808d71e4dd7cfd8003a3409ec8e8b4f5e9713a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR