meta-Phosphoric acid 33,5-36,5% stabilised ACS
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Metaphosphoric acid
Stabilisation:
Stabilised with sodium metaphosphate
33267.09
33267.22
33267.36
33267.09EA
40.6
EUR
33267.09
33267.22
33267.36
meta-Phosphoric acid 33,5-36,5% stabilised ACS
meta-Phosphoric acid
CAS No.: 37267-86-0
Molecular Formula: HPO3
Formula Weight: 79.98
Physical Form: Crystalline
Density: 2.2
Sensitivity: Hygroscopic
Appearance: White
Solubility: Decomposes in water. Soluble in alcohol
Merck Reference: 14,7345
UN No.: 3260
MDL No.: MFCD00011351
Phosphorylating agent, dehydrating agent, dental cements, laboratory reagent
Formula:
(HPO₃)n MW: 79.98 g/mol Boiling Pt: 260 Melting Pt: 21 Density: 2.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011351 CAS Number: 37267-86-0 EINECS: 253-433-4 UN: 3260 ADR: 8 III Merck Index: 14,7345 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...