Potassium hydrogen carbonate

Supplier: Roth Carl

P748.1 P748.2
ROTHP748.1EA 49.6 EUR
ROTHP748.1 ROTHP748.2
Potassium hydrogen carbonate
Potassium hydrogen carbonate
Formula: KHCO₃
Melting Pt: 292 °C
Density: 2,17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS Number: 298-14-6
EINECS: 206-059-0
Merck Index: 13,07691

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR