Digallium trisulphide ≥99.99% (metals basis)
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Gallium (III) sulphide
13387.04
13387.09
13387.02
13387.04EA
164
EUR
13387.04
13387.09
13387.02
Digallium trisulphide ≥99.99% (metals basis)
Digallium trisulphide
Purity: 99.99%
CAS No.: 12024-22-5
Molecular Formula: Ga2S3, may contain small amount of oxide
Formula Weight: 235.63
Physical Form: Powder
Density: 3.77
Melting Point: 1255°
UN No.: 3260
MDL No.: MFCD00016106
Formula:
Ga₂S₃ MW: 235,64 g/mol Melting Pt: 1255 °C Density: 3,77 Storage Temperature: Ambient |
MDL Number:
MFCD00016106 CAS Number: 12024-22-5 EINECS: 234-692-2 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...