Aluminium sulphate hydrate ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
12362.36
12362.A1
12362.A7
12362.36EA
48.6
EUR
12362.36
12362.A1
12362.A7
Aluminium sulphate hydrate ≥97%
Aluminium sulphate hydrate
Purity: 97+%
CAS No.: 17927-65-0
Molecular Formula: Al2(SO4)3.xH2O (x~14-18)
Formula Weight: 342.15(anhy)
Physical Form: Crystalline
Density: 1.61
pH: 3.5
Merck Reference: 14,366
MDL No.: MFCD00149136
Formula:
Al₂(SO₄)₃·xH₂O Density: 1,61 Storage Temperature: Ambient |
MDL Number:
MFCD00149136 CAS Number: 17927-65-0 EINECS: 233-135-0 Merck Index: 14,366 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...