Chlorobutyldimethylsilane 96%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
B 2785, Butylchlorodimethylsilane, n-Butyldimethylchlorosilane, Silan BM2, Butyldimethylchlorosilane
B23551.06
B23551.14
B23551.22
B23551.06EA
38.5
EUR
B23551.06
B23551.14
B23551.22
Chlorobutyldimethylsilane 96%
Chlorobutyldimethylsilane
Purity: 96%
CAS No.: 1000-50-6
Molecular Formula: CH3(CH2)3Si(CH3)2Cl
Formula Weight: 150.73
Flash Point: 27°(80°F)
Refractive Index: 1.4200
Boiling Point: 138°
Beilstein Reference: 1732573
UN No.: 2920
MDL No.: MFCD00015727
Formula:
C₆H₁₅ClSi MW: 150,72 g/mol Boiling Pt: 138 °C Density: 0,875 Flash Pt: 27 °C (80 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015727 CAS Number: 1000-50-6 EINECS: 423-700-1 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...