1,2-Bis(dimethylphosphino)ethane ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Ethane-1,3-diylbis(dimethylphosphine), 1,2-Bis(dimethylphosphino)ethane, Bis(1,2-dimethylphosphino)ethane, DMPE, Ethylenebis(dimethylphosphine)
B21051.03
B21051.01
B21051.03EA
175
EUR
B21051.03
B21051.01
1,2-Bis(dimethylphosphino)ethane ≥97%
1,2-Bis(dimethylphosphino)ethane
Purity: 97%
CAS No.: 23936-60-9
Molecular Formula: C6H16P2
Formula Weight: 150.14
Flash Point: -17°(1.4°F)
Refractive Index: 1.5070
Odour: Recognizable
Melting Point: 179-180°
Boiling Point: 180°
UN No.: 1993
MDL No.: MFCD00008511
Formula:
C₆H₁₆P₂ MW: 150,14 g/mol Boiling Pt: 180 °C Melting Pt: 179…180 °C Density: 0,90 Flash Pt: –17 °C (1,4 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008511 CAS Number: 23936-60-9 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...