Triphenylphosphine oxide 99%

Supplier: Thermo Scientific
Warning

140430250 140431000 140435000
ACRO140430250EA 58 EUR
ACRO140430250 ACRO140431000 ACRO140435000
Triphenylphosphine oxide 99%
Triphenylphosphine oxide
Formula: C₁₈H₁₅OP
Boiling Pt: ∼360 °C (1013 hPa)
Melting Pt: 154…158 °C
Density: 1,2 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00002080
CAS Number: 791-28-6
EINECS: 212-338-8

Order Now

Specification Test Results

Appearance White to pink-brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 154 to 158 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR