Diisopropyl sulphide ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Isopropyl sulphide
B21302.09
B21302.18
B21302.09EA
49.8
EUR
B21302.09
B21302.18
Diisopropyl sulphide ≥99%
Diisopropyl sulphide
Purity: 99%
CAS No.: 625-80-9
Molecular Formula: C6H14S
Formula Weight: 118.24
Flash Point: 7°(44°F)
Refractive Index: 1.4385
Odour: Stench
Boiling Point: 119-120°
Beilstein Reference: 1731257
UN No.: 1993
MDL No.: MFCD00008895
Formula:
C₆H₁₄S MW: 118.24 g/mol Boiling Pt: 119…120 °C Melting Pt: –69.1 °C Density: 0.814 Flash Pt: 7 °C (44 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008895 CAS Number: 625-80-9 EINECS: 210-911-7 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...