Silver tetrafluoroborate ≥99%

Supplier: Thermo Fisher Scientific
Danger

11539.09 11539.18
11539.09EA 172 EUR
11539.09 11539.18
Silver tetrafluoroborate ≥99%
Silver tetrafluoroborate

Purity: 99% CAS No.: 14104-20-2 Molecular Formula: AgBF4 Formula Weight: 194.67 Physical Form: Crystalline Sensitivity: Light Sensitive and Hygroscopic Appearance: White Melting Point: 200° dec. UN No.: 3260 MDL No.: MFCD00003408

Formula: AgBF₄
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003408
CAS Number: 14104-20-2
EINECS: 237-956-5
UN: 3260
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR