Manganese (II) acetylacetonate

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Bis(2,4-pentanedionato-O,O')manganese (II), Manganese (II) bis(2,4-pentanedionato-O,O'), Manganese(II) bis(acetylacetonate), Manganese (II) 2,4-pentanedionate

A18762.36 A18762.22 A18762.14
A18762.36EA 262 EUR
A18762.36 A18762.22 A18762.14
Manganese (II) acetylacetonate
Manganese (II) acetylacetonate

CAS No.: 14024-58-9 Molecular Formula: Mn(C5H7O2)2 Formula Weight: 253.16 Melting Point: 260-262° dec. MDL No.: MFCD00000022

Formula: C₁₀H₁₄MnO₄
MW: 253.16 g/mol
Melting Pt: 260…262 °C (dec.)
Density: 1.60
Storage Temperature: Ambient
MDL Number: MFCD00000022
CAS Number: 14024-58-9
EINECS: 237-858-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR