Tetrahydro-4H-pyran-4-one ≥98%

Supplier: Thermo Fisher Scientific
Warning

A19010.14 A19010.03 A19010.06
A19010.14EA 410 EUR
A19010.14 A19010.03 A19010.06
Tetrahydro-4H-pyran-4-one ≥98%
Tetrahydro-4H-pyran-4-one

Purity: 98% CAS No.: 29943-42-8 Molecular Formula: C5H8O2 Formula Weight: 100.12 Flash Point: 62°(144°F) Refractive Index: 1.4530 Boiling Point: 165-167° Beilstein Reference: 106463 MDL No.: MFCD00006581

Formula: C₅H₈O₂
Boiling Pt: 165…167 °C
Density: 1,084
Flash Pt: 62 °C (144 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006581
CAS Number: 29943-42-8
EINECS: 249-967-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR