4-Nitropyridine-N-oxide 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Nitropyridine N-oxide

128920050 128920250 128921000
ACRO128920050EA 28.8 EUR
ACRO128920050 ACRO128920250 ACRO128921000
4-Nitropyridine-N-oxide 97%
4-Nitropyridine N-oxide
Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
Boiling Pt: 397 °C (1013 hPa)
Melting Pt: 159…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00006206
CAS Number: 1124-33-0
EINECS: 214-395-4
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Yellow to brown Powder or crystals
Infrared spectrum Conforms
Melting point 159 to 164 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR