2,4,6-Trichloro-1,3,5-triazine 99%

Supplier: Thermo Scientific
Danger

Synonyms: Cyanuric chloride, 2,4,6-Trichloro-s-triazine

110890010 110892500 110890051
ACRO110890010EA 68.3 EUR
ACRO110890010 ACRO110892500 ACRO110890051
2,4,6-Trichloro-1,3,5-triazine 99%
2,4,6-Trichloro-1,3,5-triazine
Formula: C₃Cl₃N₃
MW: 184,41 g/mol
Boiling Pt: 190 °C (760 mmHg)
Melting Pt: 145…148 °C
Density: 1,92 g/cm³
Flash Pt: >200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006046
CAS Number: 108-77-0
EINECS: 203-614-9
UN: 2670
ADR: 8,II

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 145 to 148 °C
Titration ≥98.5 %
Anticaking agent May contain 0% to 0.5% Silica as an additive

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR