Avantor® Hichrom HI-PLUS 25, GC Columns
Supplier: Avantor
HI52-18-018-10EA 555 EUR
HI52-18-018-10 HI52-18-018-20 HI52-18-018-40 HI52-18-018-50 HI52-18-018-60 HI52-18-020-10 HI52-18-020-20 HI52-18-020-40 HI52-18-040-10 HI52-18-040-20 HI52-18-040-40 HI52-18-040-50 HI52-18-040-60 HI52-20-010-10 HI52-20-010-20 HI52-20-010-40 HI52-20-010-50 HI52-20-010-60 HI52-20-020-10 HI52-20-020-20 HI52-20-020-40 HI52-20-020-50 HI52-20-020-60 HI52-20-040-10 HI52-20-040-20 HI52-20-040-40 HI52-20-040-50 HI52-20-040-60 HI52-20-100-10 HI52-20-100-20 HI52-20-100-40 HI52-20-100-50 HI52-20-100-60 HI52-25-010-15 HI52-25-010-30 HI52-25-010-50 HI52-25-010-60 HI52-25-025-10 HI52-25-025-105 HI52-25-025-15 HI52-25-025-30 HI52-25-025-50 HI52-25-025-60 HI52-25-050-10 HI52-25-050-105 HI52-25-050-15 HI52-25-050-30 HI52-25-050-50 HI52-25-050-60 HI52-25-100-10 HI52-25-100-105 HI52-25-100-15 HI52-25-100-30 HI52-25-100-50 HI52-25-100-60 HI52-32-010-10 HI52-32-010-105 HI52-32-010-15 HI52-32-010-30 HI52-32-010-50 HI52-32-010-60 HI52-32-025-10 HI52-32-025-105 HI52-32-025-15 HI52-32-025-30 HI52-32-025-50 HI52-32-025-60 HI52-32-050-10 HI52-32-050-105 HI52-32-050-15 HI52-32-050-30 HI52-32-050-50 HI52-32-050-60 HI52-32-100-10 HI52-32-100-105 HI52-32-100-15 HI52-32-100-30 HI52-32-100-50 HI52-32-100-60 HI52-53-010-10 HI52-53-010-15 HI52-53-010-30 HI52-53-025-15 HI52-53-025-30 HI52-53-025-50 HI52-53-025-60 HI52-53-050-10 HI52-53-050-15 HI52-53-050-30 HI52-53-050-50 HI52-53-050-60 HI52-53-100-10 HI52-53-100-15 HI52-53-100-30 HI52-53-100-50 HI52-53-100-60 HI52-53-100-75 HI52-53-150-10 HI52-53-150-15 HI52-53-150-30 HI52-53-150-50 HI52-53-150-60 HI52-53-150-75 HI52-05-005-2 HI52-05-005-2.5 HI52-05-005-5 HI52-10-010-10 HI52-10-010-15 HI52-10-010-5 HI52-10-020-10 HI52-10-020-15 HI52-10-020-5 HI52-10-040-10 HI52-10-040-15 HI52-10-040-5 HI52-15-015-10 HI52-15-015-15 HI52-15-015-20 HI52-15-015-30 HI52-15-015-5 HI52-25-025-15G5 HI52-25-025-30G5 HI52-25-025-60G5 HI52-53-100-30G5
Avantor® Hichrom HI-PLUS 25, GC Columns
Chromatography Columns
Offering a range of selectivities with differing copolymer rations of PEG and methyl polysiloxane. HI-PLUS, HI-PLUS 10, HI-PLUS 25, HI-PLUS 75, HI-PLUS 90.
Max. temperature limits may change depending on the film thickness.