Cedrol
Supplier: Thermo Fisher Scientific
|
|
461420050
461420250
461421000
ACRO461420050EA
49.5
EUR
ACRO461420050
ACRO461420250
ACRO461421000
Cedrol
Cedrol
- For flavours and fragrances
Formula:
C₁₅H₂₆O Boiling Pt: 294 °C Melting Pt: 80 °C Density: 0,98 Storage Temperature: Ambient |
MDL Number:
MFCD00062952 CAS Number: 77-53-2 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | White to light yellow Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 80 to 86 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...