Tebuconazole

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-(4-Chlorophenyl)-4,4-dimethyl-3-(1H-1,2,4-triazol-1-ylmethyl)-3-pentanol

463260010 463260050
ACRO463260010EA 52.3 EUR
ACRO463260010 ACRO463260050
Tebuconazole
Tebuconazole

  • Bioactive

Formula: C₁₆H₂₂ClN₃O
MW: 307.82 g/mol
Melting Pt: 102…105 °C
Flash Pt: >100 °C
Storage Temperature: Freezer
MDL Number: MFCD02674797
CAS Number: 107534-96-3
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 100 to 107 °C
Water ≤1 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR