Zileuton

Supplier: Thermo Fisher Scientific
Warning

Synonyms: N-(1-Benzo[b]thien-2-ylethyl)-N-hydroxyurea, 1-(1-(Benzo[b]thiophen-2-yl)ethyl)-1-hydroxyurea

463300010 463300050 463300250
ACRO463300010EA 95.3 EUR
ACRO463300010 ACRO463300050 ACRO463300250
Zileuton
Zileuton

  • Bioactive

Formula: C₁₁H₁₂N₂O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00866097
CAS Number: 111406-87-2

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤1.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR