Axitinib
Supplier: Thermo Fisher Scientific
|
Warning
|
463800010
463802500
ACRO463800010EA
489
EUR
ACRO463800010
ACRO463802500
Axitinib
Axitinib
- Bioactive
Formula:
C₂₂H₁₈N₄OS Melting Pt: 226…228 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09837898 CAS Number: 319460-85-0 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | Almost white to yellow Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...