Dibutyl carbonate

Supplier: Thermo Fisher Scientific
Danger

463880050 463880250
ACRO463880050EA 70 EUR
ACRO463880050 ACRO463880250
Dibutyl carbonate
Dibutyl carbonate

  • Organic reagent

Formula: C₉H₁₈O₃
Density: 0,92
Storage Temperature: Ambient
MDL Number: MFCD00043938
CAS Number: 542-52-9

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4140 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR