Adapalene
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoic Acid, 6-(3-(Adamantan-1-yl)-4-methoxyphenyl)-2-naphthoic acid
464500010
464500050
ACRO464500010EA
145
EUR
ACRO464500010
ACRO464500050
Adapalene
Adapalene
- Bioactive
Formula:
C₂₈H₂₈O₃ MW: 412,53 g/mol Melting Pt: 324 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03106112 CAS Number: 106685-40-9 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % (105°C, 4 h) |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...