Dapagliflozin
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(2S,3R,4R,5S,6R)-2-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
464620010
464620050
ACRO464620010EA
148
EUR
ACRO464620010
ACRO464620050
Dapagliflozin
Dapagliflozin
- Bioactive
Formula:
C₂₁H₂₅ClO₆ MW: 408,87 g/mol Melting Pt: 65…70 °C Storage Temperature: Freezer |
MDL Number:
MFCD13182359 CAS Number: 461432-26-8 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤2 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...