Sesamin
Supplier: Thermo Scientific
Synonyms:
(1S,3aR,4S,6aR)-1,4-Bis(benzo[d][1,3]dioxol-5-yl)hexahydrofuro[3,4-c]furan
464890010
464892500
ACRO464890010EA
568
EUR
ACRO464890010
ACRO464892500
Sesamin
Sesamin
- Bioactive
Formula:
C₂₀H₁₈O₆ MW: 354,36 g/mol Melting Pt: 123 °C Storage Temperature: Freezer |
MDL Number:
MFCD00216134 CAS Number: 607-80-7 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 121 to 125 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...