Ceftizoxime

Supplier: Thermo Scientific

Synonyms: (6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

465110010 465110050
ACRO465110010EA 91 EUR
ACRO465110010 ACRO465110050
Ceftizoxime
Ceftizoxime

  • Bioactive

Formula: C₁₃H₁₃N₅O₅S₂
MW: 383,40 g/mol
Melting Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00072000
CAS Number: 68401-81-0

Order Now

Specification Test Results

Appearance White to almost white to light yellow Powder
Infrared spectrum Conforms
Water ≤2 %
HPLC ≥98.5 %
Residue after ign. (sulfated) ≤0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR