(S,R)-Noscapine
Supplier: Thermo Fisher Scientific
|
Warning
|
462710050
462710250
ACRO462710050EA
71
EUR
ACRO462710050
ACRO462710250
(S,R)-Noscapine
(S,R)-Noscapine
- Bioactive
Formula:
C₂₂H₂₃NO₇ MW: 413.42 g/mol Melting Pt: 174…177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00069316 CAS Number: 128-62-1 |
Specification Test Results
Appearance | Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 174 to 177 °C |
Loss on drying | ≤1 % (105 °C) |
HPLC | ≥98.5 % |
Residue after ignition | ≤0.1 % |
Specific optical rotation | +42° to +48° (20 °C, 589 nm) (2 % in 0.1M HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...