Silibinin

Supplier: Thermo Scientific
Warning

Synonyms: Silybin A, (2R,3R)-3,5,7-Trihydroxy-2-((2R,3R)-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)chroman-4-one

463030010 463030050 463030250
ACRO463030010EA 43.2 EUR
ACRO463030010 ACRO463030050 ACRO463030250
Silibinin
Silibinin

  • Bioactive

Formula: C₂₅H₂₂O₁₀
MW: 482,44 g/mol
Melting Pt: 164…174 °C
Storage Temperature: Freezer
MDL Number: MFCD03225424
CAS Number: 22888-70-6

Order Now

Specification Test Results

Appearance Almost white to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤4 %
HPLC ≥97.5 % (mixture of A and B)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR