Bifenox
Supplier: Thermo Fisher Scientific
|
Warning
|
464600010
464602500
ACRO464600010EA
409
EUR
ACRO464600010
ACRO464602500
Bifenox
Bifenox
- Bioactive
Formula:
C₁₄H₉Cl₂NO₅ Melting Pt: 84…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055314 CAS Number: 42576-02-3 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | White to yellow to light beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 83 to 89 °C |
Water | ≤1 % |
HPLC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...