PPDT2FBT (PCE9.3), Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific
Synonyms:
Poly[(2,5-bis(2-hexyldecyloxy)phenylene)-alt-(5,6-difluoro-4,7-di(thiophen-2-yl)benzo[c]-[1,2,5]thiadiazole)]
H66399.MC
H66399.MD
H66399.ME
H66399.MCEA
565
EUR
H66399.MC
H66399.MD
H66399.ME
PPDT2FBT (PCE9.3), Organic Photovoltaics (OPV) reagent
PPDT2FBT (PCE9.3)
Formula:
(C₅₂H₇₂F₂N₂O₂S₃)n |
CAS Number:
1620673-07-5 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1,78 ± 0,02 eV |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...