PBDB-T-2Cl, Organic Photovoltaics (OPV) reagent

Supplier: Thermo Fisher Scientific

H66867.MC H66867.MD H66867.ME
H66867.MCEA 777 EUR
H66867.MC H66867.MD H66867.ME
PBDB-T-2Cl, Organic Photovoltaics (OPV) reagent
PBDB-T-2Cl
Formula: (C₆₈H₇₆Cl₂O₂S₈)n
CAS Number: 2239295-71-5

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy Tbd


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR