PDPPTT, Organic Photovoltaics (OPV) reagent

Supplier: Thermo Scientific

Synonyms: Poly[2,5-(2-decyltetradecyl)-3,6-diketopyrrolopyrrole-alt-5,5-(2,5- di(thien-2- yl)thieno[3,2-b]thiophene)]

H66319.MC H66319.MD H66319.ME
H66319.MCEA 708 EUR
H66319.MC H66319.MD H66319.ME
PDPPTT, Organic Photovoltaics (OPV) reagent
PDPPTT
Formula: (C₆₈H₁₀₄N₂O₂S₄)n
CAS Number: 1260685-66-2

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1,40 ± 0,02 eV

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR