poly(3-hexylthiophene-2,5-diyl), Organic Photovoltaics (OPV) reagent

Supplier: Thermo Fisher Scientific

Synonyms: P3HT, Poly(3-hexylthiophene), Poly[3-hexylthiophene-2,5-diyl]

H66726.ME H66726.03
H66726.MEEA 617 EUR
H66726.ME H66726.03
poly(3-hexylthiophene-2,5-diyl), Organic Photovoltaics (OPV) reagent
Poly(3-hexylthiophene-2,5-diyl)
MDL Number: MFCD00217686
CAS Number: 104934-50-1

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy T.b.d.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR