Thymoquinone ≥98%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
|
Warning
|
Synonyms:
p-Mentha-3,6-diene-2,5-dione, 2-Isopropyl-5-methylcyclohexa-2,5-diene-1,4-dione, 5-Isopropyl-2-methyl-1,4-benzoquinone
M-Clarity™ quality level = MQ100
Thymoquinone has been used as an inhibitor of Escherichia coli ATP synthase and cell growth. It is also used as a drug against lower limb ischemia-reperfusion injury of sciatic nerve and femoral muscle. Thymoquinone is employed in preclinical studies as a neuroprotective and anticonvulsant agent.
Thymoquinone (TQ), also known as 2-isopropyl-5-methyl-1, 4-benzoquinone, is present in a crystalline triclinic form. It is the bioactive constituent of the volatile oil of black seed. TQ exhibits anti-inflammatory, antioxidant and anti-neoplastic effects both in vitro and in vivo.
Formula:
C₁₀H₁₂O₂ MW: 164,20 g/mol Boiling Pt: 230,..232 °C (lit.) Melting Pt: 45,..47 °C (lit.) Flash Pt: 104 °C (closed cup) Storage Temperature: Room Temperature |
MDL Number:
MFCD00001602 CAS Number: 490-91-5 EINECS: 207-721-1 |
Specification Test Results
Appearance (Colour) | White to Dark Yellow and Faint Orange to Orange and Faint Brown to Brown and Orange-Brown and Brown-Orange |
Appearance (Form) | Powder or Crystal or Chunk(s) |
Infrared spectrum | Conforms to Structure |
Purity (GC) | ≥98.0% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...