Disinfectant wipes, Azowipe™
Supplier: Vernacare
Azowipe™
VERO00720EA 0 EUR
VERO00720
Disinfectant wipes, Azowipe™
Wipes Disposable Wipes and Towels Disinfectant Wipes
Sterile triple bagged wipes; Azowipe is convenient for use in all clinical environments.
  • Saturated with 70% solution w/w of isopropyl alcohol BP
  • Hard surface cleaning wipes effective against MRSA, E.coli and various other micro-organisms
Caution: Use biocides safely. Always read the label and product information before use.
*The product(s) on this page are outside the scope of the EU Directive 2019/904