Alternariol 99%

Supplier: Apollo Scientific
Danger

Synonyms: 3,7,9-Trihydroxy-1-methyl-6H-dibenzo(b,d)pyran-6-one

BIA2301-1MG BIA2301-5MG
APOSBIA2301-1MGEA 171 EUR
APOSBIA2301-1MG APOSBIA2301-5MG
Alternariol 99%
Alternariol

From Alternaria, mycotoxin, food contaminant

Formula: C₁₄H₁₀O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00133068
CAS Number: 641-38-3
UN: 3462
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR