Palatinose

Supplier: Apollo Scientific
Warning

Synonyms: Isomaltulose, 6-O-alpha-D-Glucopyranosyl-D-fructose, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(((2R,3S,4S,5R)-3,4,5-trihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)methoxy)tetrahydro-2H-pyran-3,4,5-triol

BIB6254-1KG BIB6254-250G BIB6254-5KG
APOSBIB6254-1KGEA 255 EUR
APOSBIB6254-1KG APOSBIB6254-250G APOSBIB6254-5KG
Palatinose
Palatinose
Formula: C₁₂H₂₂O₁₁
MW: 342,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00076094
CAS Number: 13718-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR