1-Azido-1-deoxy-β-D-glucopyranoside ≥98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
β-D-Glucopyranosyl azide
BICL4025-1G
BICL4025-500MG
BICL4025-100MG
APOSBICL4025-1GEA
945
EUR
APOSBICL4025-1G
APOSBICL4025-500MG
APOSBICL4025-100MG
1-Azido-1-deoxy-β-D-glucopyranoside ≥98%
1-Azido-1-deoxy-β-D-glucopyranoside
Formula:
C₆H₁₁N₃O₅ MW: 205,169 g/mol Melting Pt: 150 °C (dec.) Storage Temperature: Freezer |
MDL Number:
MFCD03701503 CAS Number: 20379-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...