3,4-Dideoxyglucosone-3-ene ≥98%

Supplier: Apollo Scientific
Warning

Synonyms: 3,4-DGE

BICL4066-1MG BICL4066-5MG BICL4066-2MG
APOSBICL4066-1MGEA 517 EUR
APOSBICL4066-1MG APOSBICL4066-5MG APOSBICL4066-2MG
3,4-Dideoxyglucosone-3-ene ≥98%
3,4-Dideoxyglucosone-3-ene
Formula: C₆H₈O₄
MW: 144.125 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02683312
CAS Number: 252006-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR