Furosemide-acyl-β-D-glucuronide ≥92.3%

Supplier: Apollo Scientific

BICL4097-1MG BICL4097-2MG
APOSBICL4097-1MGEA 475 EUR
APOSBICL4097-1MG APOSBICL4097-2MG
Furosemide-acyl-β-D-glucuronide ≥92.3%
Furosemide-acyl-β-D-glucuronide
Storage Temperature: Ambient
CAS Number: 72967-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR