Febuxostat-acyl-β-D-glucuronide ≥96%

Supplier: Apollo Scientific

BICL4103-1MG BICL4103-5MG BICL4103-2MG
APOSBICL4103-1MGEA 337 EUR
APOSBICL4103-1MG APOSBICL4103-5MG APOSBICL4103-2MG
Febuxostat-acyl-β-D-glucuronide ≥96%
Febuxostat-acyl-β-D-glucuronide
Storage Temperature: Ambient
CAS Number: 1351692-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR