1-O-Galloyl β-D-glucopyranoside ≥98%

Supplier: Apollo Scientific
Warning

Synonyms: β-D-Glucogallin, 1-O-(3,4,5-Trihydroxybenzoyl)-β-D-glucopyranoside, β-D-Glucose gallate

BICL4108-250MG BICL4108-50MG BICL4108-100MG
APOSBICL4108-250MGEA 1390 EUR
APOSBICL4108-250MG APOSBICL4108-50MG APOSBICL4108-100MG
1-O-Galloyl β-D-glucopyranoside ≥98%
1-O-Galloyl β-D-glucopyranoside
Formula: C₁₃H₁₆O₁₀
MW: 332,26 g/mol
Melting Pt: 200…205 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD11975493
CAS Number: 13405-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR