Mycophenolic acid β-D-glucopyranoside ≥98%
Supplier: Apollo Scientific
Synonyms:
Mycophenolic acid-phenolic-β-D-glucoside
BICL4133-1MG
BICL4133-5MG
BICL4133-2MG
APOSBICL4133-1MGEA
323
EUR
APOSBICL4133-1MG
APOSBICL4133-5MG
APOSBICL4133-2MG
Mycophenolic acid β-D-glucopyranoside ≥98%
Mycophenolic acid β-D-glucopyranoside
Storage Temperature:
Ambient |
CAS Number:
55533-52-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...