4-Methylphenyl 2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside ≥99%

Supplier: Apollo Scientific

Synonyms: Tolyl 2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside, Thiocresyl 2,3,4,6-tetra-O-acetyl-β-D-glucopyranoside

BICL4144-10G BICL4144-2G BICL4144-5G
APOSBICL4144-10GEA 1120 EUR
APOSBICL4144-10G APOSBICL4144-2G APOSBICL4144-5G
4-Methylphenyl 2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside ≥99%
4-Methylphenyl 2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside
Formula: C₂₁H₂₆O₉S
MW: 454,4908 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13182351
CAS Number: 28244-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR