4-Methylphenyl 6-O-acetyl-2,3,4-tri-O-benzyl-1-thio-α-D-glucopyranoside ≥99%
Supplier: Apollo Scientific
Synonyms:
Tolyl 6-O-acetyl-2,3,4-tri-O-benzyl-1-thio-α-D-glucopyranoside, Thiocresyl 6-O-acetyl-2,3,4-tri-O-benzyl-α-D-glucopyranoside
BICL4146-1G
BICL4146-2G
BICL4146-500MG
APOSBICL4146-1GEA
685
EUR
APOSBICL4146-1G
APOSBICL4146-2G
APOSBICL4146-500MG
4-Methylphenyl 6-O-acetyl-2,3,4-tri-O-benzyl-1-thio-α-D-glucopyranoside ≥99%
4-Methylphenyl 6-O-acetyl-2,3,4-tri-O-benzyl-1-thio-α-D-glucopyranoside
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...