Methyl 1,2,3,4-tetra-O-acetyl-α-L-idopyranosyluronate ≥98%
Supplier: Apollo Scientific
Synonyms:
1,2,3,4-Tetra-O-acetyl-α-L-idopyranuronic acid methyl ester
BICL4163-100MG
BICL4163-25MG
BICL4163-50MG
APOSBICL4163-100MGEA
1190
EUR
APOSBICL4163-100MG
APOSBICL4163-25MG
APOSBICL4163-50MG
Methyl 1,2,3,4-tetra-O-acetyl-α-L-idopyranosyluronate ≥98%
Methyl 1,2,3,4-tetra-O-acetyl-α-L-idopyranosyluronate
Storage Temperature:
Ambient |
CAS Number:
108032-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...